Assay Details
| Gene Symbol: Mapk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081567.1 | NP_001075036.1 | 3 - 4 | 634 | 100 | |
| NM_009158.2 | NP_033184.2 | 3 - 4 | 634 | 100 | ||
| GenBank mRNA | AB096076.1 | - | 1 - 2 | 68 | 100 | |
| AB096077.1 | - | 1 - 2 | 68 | 100 | ||
| AB096078.1 | - | 1 - 2 | 68 | 100 | ||
| AB096079.1 | - | 1 - 2 | 68 | 100 | ||
| AK042368.1 | - | 2 - 3 | 199 | 100 | ||
| AK082242.1 | - | 3 - 4 | 637 | 100 | ||
| AK163206.1 | - | 3 - 4 | 585 | 100 | ||
| BC046625.1 | - | 3 - 4 | 573 | 6494163 | 100 | |
| L35236.1 | - | 3 - 4 | 377 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC046625.1 transcripts.
Set Membership: |
|

Mapk10
Genomic Map