Assay Details
| Gene Symbol: Adrbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285806.1 | NP_001272735.1 | 17 - 18 | 1593 | 80 | |
| NM_177078.4 | NP_796052.2 | 17 - 18 | 1554 | 80 | ||
| GenBank mRNA | AK076533.1 | - | 17 - 18 | 1452 | 80 | |
| AK134672.1 | - | 17 - 18 | 1531 | 80 | ||
| AK141186.1 | - | 17 - 18 | 1573 | 80 | ||
More Information
Set Membership: |
|

Adrbk2
Genomic Map