Assay Details
| Gene Symbol: Wbscr22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025375.3 | NP_079651.2 | 4 - 5 | 442 | 85 | |
| GenBank mRNA | AF412035.1 | - | 3 - 4 | 214 | 85 | |
| AK002497.1 | - | 2 - 3 | 536 | 85 | ||
| AK003386.1 | - | 3 - 4 | 215 | 85 | ||
| AK008966.1 | - | 1 - 2 | 47 | 85 | ||
| AK011005.1 | - | 3 - 4 | 222 | 85 | ||
| AK151986.1 | - | 3 - 4 | 223 | 85 | ||
| AK178657.1 | - | 1 - 2 | 109 | 85 | ||
| BC093484.1 | - | 3 - 4 | 192 | 6735279 | 85 | |
| BC117986.1 | - | 4 - 5 | 442 | 40102067 | 85 | |
| BC117987.1 | - | 4 - 5 | 442 | 40102068 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wbscr22
Genomic Map