Assay Details
| Gene Symbol: Wbscr22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025375.3 | NP_079651.2 | 8 - 9 | 772 | 55 | |
| GenBank mRNA | AF412035.1 | - | 7 - 8 | 544 | 55 | |
| AK002497.1 | - | 6 - 7 | 866 | 55 | ||
| AK008966.1 | - | 5 - 6 | 377 | 55 | ||
| AK011005.1 | - | 7 - 8 | 552 | 55 | ||
| AK151986.1 | - | 7 - 8 | 553 | 55 | ||
| BC093484.1 | - | 7 - 8 | 522 | 6735279 | 55 | |
| BC117986.1 | - | 8 - 9 | 772 | 40102067 | 55 | |
| BC117987.1 | - | 8 - 9 | 772 | 40102068 | 55 | |
More Information
Set Membership: |
|

Wbscr22
Genomic Map