Assay Details
| Gene Symbol: Eif4h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033561.1 | NP_291039.1 | 5 - 6 | 502 | 141 | |
| GenBank mRNA | AK045004.1 | - | 5 - 6 | 504 | 141 | |
| AK088818.1 | - | 5 - 6 | 514 | 141 | ||
| AK132034.1 | - | 5 - 6 | 522 | 141 | ||
| AK135390.1 | - | 5 - 6 | 503 | 141 | ||
| AK149841.1 | - | 5 - 6 | 488 | 141 | ||
| AK150088.1 | - | 5 - 6 | 511 | 141 | ||
| AK166567.1 | - | 5 - 6 | 503 | 141 | ||
| AK169312.1 | - | 5 - 6 | 485 | 141 | ||
| AK169353.1 | - | 5 - 6 | 503 | 141 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif4h
Genomic Map