Assay Details
| Gene Symbol: Pptc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177242.4 | NP_796216.2 | 4 - 5 | 1013 | 66 | |
| GenBank mRNA | AK078914.1 | - | 4 - 5 | 1021 | 66 | |
| AK083481.1 | - | 4 - 5 | 1030 | 66 | ||
| AK135118.1 | - | 4 - 5 | 1047 | 66 | ||
| AK162261.1 | - | 4 - 5 | 577 | 66 | ||
| BC038622.1 | - | 4 - 5 | 653 | 66 | ||
| BC068149.1 | - | 4 - 5 | 1013 | 30537082 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pptc7
Genomic Map