Assay Details
| Gene Symbol: Klk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174865.1 | NP_777354.1 | 3 - 4 | 475 | 55 | |
| GenBank mRNA | BC106965.1 | - | 3 - 4 | 475 | 40039744 | 55 |
| BC106966.2 | - | 3 - 4 | 472 | 55 | ||
| BC107012.1 | - | 3 - 4 | 475 | 40047687 | 55 | |
More Information
Set Membership: |
|

Klk15
Genomic Map