| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900003407.mm.1 |
NM_146096.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003407.mm.1 |
NM_146096.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003407.mm.1 |
TC1900000344.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003409.mm.1 |
NM_146096.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003409.mm.1 |
NM_146096.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003409.mm.1 |
TC1900000344.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003405.mm.1 |
NM_146096.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003405.mm.1 |
NM_146096.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003405.mm.1 |
TC1900000344.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003406.mm.1 |
NM_146096.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003406.mm.1 |
NM_146096.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003406.mm.1 |
TC1900000344.mm.1 |
MTA TC |