Assay Details
| Gene Symbol: Parp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163575.1 | NP_001157047.1 | 1 - 2 | 92 | 88 | |
| GenBank mRNA | AK028373.1 | - | 1 - 2 | 40 | 88 | |
| AK035309.1 | - | 1 - 2 | 86 | 88 | ||
| AK146336.1 | - | 1 - 2 | 93 | 88 | ||
| BC024074.1 | - | 1 - 2 | 49 | 88 | ||
| BY742887.1 | - | 1 - 2 | 93 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028373.1, BC024074.1, NM_001163575.1 transcripts.
Set Membership: |
|

Parp10
Genomic Map