Assay Details
| Gene Symbol: Parp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163575.1 | NP_001157047.1 | 2 - 3 | 274 | 90 | |
| NM_001163576.1 | NP_001157048.1 | 1 - 2 | 378 | 90 | ||
| GenBank mRNA | AK028373.1 | - | 2 - 3 | 222 | 90 | |
| AK035309.1 | - | 2 - 3 | 268 | 90 | ||
| AK046755.1 | - | 1 - 2 | 359 | 90 | ||
| AK146336.1 | - | 2 - 3 | 275 | 90 | ||
| AK153205.1 | - | 1 - 2 | 115 | 90 | ||
| AK166291.1 | - | 2 - 3 | 225 | 90 | ||
| BC024074.1 | - | 2 - 3 | 231 | 90 | ||
| BY742887.1 | - | 2 - 3 | 275 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK046755.1 transcripts.
Set Membership: |
|

Parp10
Genomic Map