Assay Details
| Gene Symbol: Parp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163575.1 | NP_001157047.1 | 4 - 5 | 763 | 75 | |
| NM_001163576.1 | NP_001157048.1 | 3 - 4 | 867 | 75 | ||
| GenBank mRNA | AK028373.1 | - | 4 - 5 | 711 | 75 | |
| AK046755.1 | - | 3 - 4 | 848 | 75 | ||
| AK146336.1 | - | 4 - 5 | 764 | 75 | ||
| AK153205.1 | - | 3 - 4 | 604 | 75 | ||
| AK166291.1 | - | 4 - 5 | 714 | 75 | ||
| BC024074.1 | - | 4 - 5 | 720 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK046755.1 transcripts.
Set Membership: |
|

Parp10
Genomic Map