Assay Details
| Gene Symbol: Parp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163575.1 | NP_001157047.1 | 5 - 6 | 1431 | 89 | |
| NM_001163576.1 | NP_001157048.1 | 4 - 5 | 1535 | 89 | ||
| GenBank mRNA | AK028373.1 | - | 5 - 6 | 1379 | 89 | |
| AK035309.1 | - | 4 - 5 | 1505 | 89 | ||
| AK046755.1 | - | 4 - 5 | 1516 | 89 | ||
| AK146336.1 | - | 5 - 6 | 1432 | 89 | ||
| AK153205.1 | - | 4 - 5 | 1272 | 89 | ||
| AK166291.1 | - | 5 - 6 | 1382 | 89 | ||
| BC023056.1 | - | 1 - 2 | 268 | 89 | ||
| BC024074.1 | - | 5 - 6 | 1388 | 89 | ||
| BC025608.1 | - | 1 - 2 | 413 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Parp10
Genomic Map