| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500013492.mm.1 |
NM_001163575.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013492.mm.1 |
NM_001163575.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013492.mm.1 |
NM_001163576.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013492.mm.1 |
NM_001163576.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013492.mm.1 |
TC1500002347.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500019352.mm.1 |
NM_001163575.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500019352.mm.1 |
NM_001163575.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500019352.mm.1 |
NM_001163576.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500019352.mm.1 |
NM_001163576.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500019352.mm.1 |
TC1500002347.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500013491.mm.1 |
NM_001163575.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013491.mm.1 |
NM_001163575.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013491.mm.1 |
NM_001163576.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013491.mm.1 |
NM_001163576.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013491.mm.1 |
TC1500002347.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500013495.mm.1 |
NM_001163575.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013495.mm.1 |
NM_001163575.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013495.mm.1 |
NM_001163576.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013495.mm.1 |
NM_001163576.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013495.mm.1 |
TC1500002347.mm.1 |
MTA TC |