Assay Details
| Gene Symbol: Gusb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010368.1 | NP_034498.1 | 7 - 8 | 1241 | 73 | |
| GenBank mRNA | AK041058.1 | - | 6 - 7 | 1060 | 73 | |
| AK136519.1 | - | 7 - 8 | 1298 | 73 | ||
| AK146247.1 | - | 7 - 8 | 1462 | 73 | ||
| AK149814.1 | - | 7 - 8 | 1270 | 73 | ||
| AK150048.1 | - | 7 - 8 | 1242 | 73 | ||
| AK151802.1 | - | 7 - 8 | 1242 | 73 | ||
| AK152206.1 | - | 7 - 8 | 1242 | 73 | ||
| AK159163.1 | - | 7 - 8 | 1242 | 73 | ||
| AK159526.1 | - | 7 - 8 | 1242 | 73 | ||
| AK159564.1 | - | 7 - 8 | 1233 | 73 | ||
| AK159804.1 | - | 7 - 8 | 1242 | 73 | ||
| AK162436.1 | - | 7 - 8 | 1242 | 73 | ||
| BC071226.1 | - | 7 - 8 | 1262 | 5325429 | 73 | |
| J03047.1 | - | 7 - 8 | 1241 | 73 | ||
| M19279.1 | - | 7 - 8 | 1241 | 73 | ||
| M28540.1 | - | 7 - 8 | 1241 | 73 | ||
| M28541.1 | - | 7 - 8 | 1241 | 73 | ||
| M63836.1 | - | 7 - 8 | 1241 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gusb
Genomic Map