Assay Details
| Gene Symbol: Sumf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026445.2 | NP_080721.1 | 2 - 3 | 250 | 74 | |
| GenBank mRNA | AK013017.1 | - | 2 - 3 | 230 | 74 | |
| AK076022.1 | - | 2 - 3 | 256 | 74 | ||
| AK138300.1 | - | 2 - 3 | 241 | 74 | ||
| AK140169.1 | - | 2 - 3 | 221 | 74 | ||
| AK157784.1 | - | 2 - 3 | 251 | 74 | ||
| AK166216.1 | - | 2 - 3 | 229 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sumf2
Genomic Map