Assay Details
| Gene Symbol: Sumf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026445.2 | NP_080721.1 | 3 - 4 | 361 | 80 | |
| GenBank mRNA | AK013017.1 | - | 3 - 4 | 341 | 80 | |
| AK076022.1 | - | 3 - 4 | 367 | 80 | ||
| AK138300.1 | - | 3 - 4 | 352 | 80 | ||
| AK140169.1 | - | 3 - 4 | 332 | 80 | ||
| AK157784.1 | - | 3 - 4 | 362 | 80 | ||
| AK166216.1 | - | 3 - 4 | 340 | 80 | ||
| BC104404.1 | - | 1 - 2 | 470 | 80 | ||
| BC104405.1 | - | 1 - 2 | 470 | 80 | ||
| BC119479.1 | - | 1 - 2 | 470 | 80 | ||
| BC119480.2 | - | 1 - 2 | 470 | 40050522 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC119480.2 transcripts.
Set Membership: |
|

Sumf2
Genomic Map