Assay Details
| Gene Symbol: Sumf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026445.2 | NP_080721.1 | 6 - 7 | 621 | 59 | |
| GenBank mRNA | AK013017.1 | - | 6 - 7 | 601 | 59 | |
| AK076022.1 | - | 6 - 7 | 627 | 59 | ||
| AK138300.1 | - | 6 - 7 | 612 | 59 | ||
| AK140169.1 | - | 7 - 8 | 636 | 59 | ||
| AK157784.1 | - | 6 - 7 | 622 | 59 | ||
| BC119480.2 | - | 4 - 5 | 730 | 40050522 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sumf2
Genomic Map