Assay Details
| Gene Symbol: Cd209b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037800.3 | NP_001032889.3 | 5 - 6 | 433 | 70 | |
| NM_001287211.1 | NP_001274140.1 | 5 - 6 | 433 | 70 | ||
| NM_026972.5 | NP_081248.4 | 6 - 7 | 523 | 70 | ||
| NR_104430.2 | 6 - 7 | 523 | 70 | |||
| GenBank mRNA | AF373409.1 | - | 6 - 7 | 501 | 70 | |
| AF374471.1 | - | 5 - 6 | 390 | 70 | ||
| AF422108.1 | - | 6 - 7 | 496 | 70 | ||
| AF424790.1 | - | 6 - 7 | 495 | 70 | ||
| AF424792.1 | - | 6 - 7 | 495 | 70 | ||
| AF424793.1 | - | 6 - 7 | 495 | 70 | ||
| AF424794.1 | - | 6 - 7 | 515 | 70 | ||
| AF424795.1 | - | 6 - 7 | 512 | 70 | ||
| AF424796.1 | - | 6 - 7 | 506 | 70 | ||
| AF424797.1 | - | 5 - 6 | 405 | 70 | ||
| AF424798.1 | - | 5 - 6 | 425 | 70 | ||
| AF424799.1 | - | 5 - 6 | 405 | 70 | ||
| AF424803.1 | - | 6 - 7 | 531 | 70 | ||
| AK007656.1 | - | 5 - 6 | 439 | 70 | ||
| AK138999.1 | - | 6 - 7 | 497 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd209b
Genomic Map