Assay Details
| Gene Symbol: Gsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010344.4 | NP_034474.4 | 11 - 12 | 1524 | 112 | |
| GenBank mRNA | AK019177.1 | - | 11 - 12 | 1183 | 112 | |
| AK040136.1 | - | 11 - 12 | 1233 | 112 | ||
| AK084328.1 | - | 11 - 12 | 1154 | 112 | ||
| AK151063.1 | - | 11 - 12 | 1170 | 112 | ||
| AK151247.1 | - | 11 - 12 | 1170 | 112 | ||
| AK151674.1 | - | 11 - 12 | 1082 | 112 | ||
| AK152873.1 | - | 11 - 12 | 1173 | 112 | ||
| AK159218.1 | - | 11 - 12 | 1173 | 112 | ||
| AK159678.1 | - | 11 - 12 | 1196 | 112 | ||
| AK167393.1 | - | 11 - 12 | 1154 | 111 | ||
| BC006966.1 | - | 8 - 9 | 849 | 112 | ||
| BC056357.1 | - | 11 - 12 | 1524 | 6813295 | 112 | |
| BC057325.1 | - | 11 - 12 | 1524 | 6813295 | 112 | |
| X76341.2 | - | 11 - 12 | 1222 | 112 | ||
More Information
Set Membership: |
|

Gsr
Genomic Map