Assay Details
| Gene Symbol: Gsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010344.4 | NP_034474.4 | 7 - 8 | 1035 | 86 | |
| GenBank mRNA | AK040136.1 | - | 7 - 8 | 744 | 86 | |
| AK084328.1 | - | 7 - 8 | 665 | 86 | ||
| AK151063.1 | - | 7 - 8 | 681 | 86 | ||
| AK151247.1 | - | 7 - 8 | 681 | 86 | ||
| AK151674.1 | - | 7 - 8 | 593 | 86 | ||
| AK159218.1 | - | 7 - 8 | 684 | 86 | ||
| AK159678.1 | - | 7 - 8 | 707 | 86 | ||
| AK167393.1 | - | 7 - 8 | 666 | 86 | ||
| BC006966.1 | - | 4 - 5 | 360 | 86 | ||
| BC056357.1 | - | 7 - 8 | 1035 | 6813295 | 86 | |
| BC057325.1 | - | 7 - 8 | 1035 | 6813295 | 86 | |
| BQ178602.1 | - | 4 - 5 | 345 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gsr
Genomic Map