Assay Details
| Gene Symbol: Gsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010344.4 | NP_034474.4 | 10 - 11 | 1391 | 73 | |
| GenBank mRNA | AK019177.1 | - | 10 - 11 | 1050 | 73 | |
| AK040136.1 | - | 10 - 11 | 1100 | 73 | ||
| AK084328.1 | - | 10 - 11 | 1021 | 73 | ||
| AK151063.1 | - | 10 - 11 | 1037 | 73 | ||
| AK151247.1 | - | 10 - 11 | 1037 | 73 | ||
| AK151674.1 | - | 10 - 11 | 949 | 73 | ||
| AK152873.1 | - | 10 - 11 | 1040 | 73 | ||
| AK159218.1 | - | 10 - 11 | 1040 | 73 | ||
| AK159678.1 | - | 10 - 11 | 1063 | 73 | ||
| AK167393.1 | - | 10 - 11 | 1022 | 73 | ||
| BC006966.1 | - | 7 - 8 | 716 | 73 | ||
| BC056357.1 | - | 10 - 11 | 1391 | 6813295 | 73 | |
| BC057325.1 | - | 10 - 11 | 1391 | 6813295 | 73 | |
| BQ178602.1 | - | 7 - 8 | 701 | 73 | ||
| X76341.2 | - | 10 - 11 | 1089 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gsr
Genomic Map