Assay Details
| Gene Symbol: Tex15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031374.2 | NP_113551.2 | 2 - 3 | 7089 | 151 | |
| GenBank mRNA | AF285589.1 | - | 2 - 3 | 7089 | 151 | |
| AK008726.1 | - | 2 - 3 | 479 | 151 | ||
| BC063106.1 | - | 9 - 10 | 1310 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tex15
Genomic Map