Assay Details
| Gene Symbol: Rbpms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042674.2 | NP_001036139.1 | 3 - 4 | 502 | 80 | |
| NM_001042675.2 | NP_001036140.1 | 3 - 4 | 502 | 80 | ||
| NM_001286168.1 | NP_001273097.1 | 3 - 4 | 502 | 80 | ||
| NM_019733.3 | NP_062707.1 | 3 - 4 | 502 | 80 | ||
| GenBank mRNA | AF148511.1 | - | 3 - 4 | 451 | 80 | |
| AK012265.1 | - | 3 - 4 | 445 | 80 | ||
| AK012586.1 | - | 3 - 4 | 445 | 80 | ||
| AK041118.1 | - | 3 - 4 | 446 | 80 | ||
| AK079238.1 | - | 3 - 4 | 445 | 80 | ||
| BB637557.1 | - | 3 - 4 | 446 | 80 | ||
| BC030397.1 | - | 3 - 4 | 463 | 4225078 | 80 | |
| CK030989.1 | - | 3 - 4 | 480 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbpms
Genomic Map