Assay Details
| Gene Symbol: Rbpms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019733.3 | NP_062707.1 | 6 - 7 | 851 | 100 | |
| GenBank mRNA | AF148511.1 | - | 6 - 7 | 800 | 100 | |
| BC030397.1 | - | 6 - 7 | 812 | 4225078 | 100 | |
| BY708385.1 | - | 3 - 4 | 345 | 100 | ||
More Information
Set Membership: |
|

Rbpms
Genomic Map