Assay Details
| Gene Symbol: Gtf2e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167921.1 | NP_001161393.1 | 2 - 3 | 250 | 77 | |
| NM_001167922.1 | NP_001161394.1 | 2 - 3 | 352 | 77 | ||
| NM_026584.3 | NP_080860.3 | 2 - 3 | 238 | 77 | ||
| GenBank mRNA | AK007492.1 | - | 1 - 2 | 195 | 77 | |
| AK021183.1 | - | 2 - 3 | 203 | 77 | ||
| AK088342.1 | - | 2 - 3 | 263 | 77 | ||
| AK146803.1 | - | 2 - 3 | 225 | 77 | ||
| AK150415.1 | - | 2 - 3 | 196 | 77 | ||
| AK153370.1 | - | 2 - 3 | 196 | 77 | ||
| AK158783.1 | - | 2 - 3 | 332 | 77 | ||
| BC020016.1 | - | 2 - 3 | 218 | 3501113 | 77 | |
| BY256665.1 | - | 2 - 3 | 239 | 77 | ||
| CX239577.1 | - | 2 - 3 | 250 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2e2
Genomic Map