Assay Details
| Gene Symbol: Btrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037758.2 | NP_001032847.2 | 7 - 8 | 944 | 82 | |
| NM_001286465.1 | NP_001273394.1 | 7 - 8 | 944 | 82 | ||
| NM_001286466.1 | NP_001273395.1 | 6 - 7 | 712 | 82 | ||
| NM_009771.3 | NP_033901.1 | 6 - 7 | 836 | 82 | ||
| GenBank mRNA | AF081887.1 | - | 6 - 7 | 821 | 82 | |
| AF099932.1 | - | 6 - 7 | 731 | 82 | ||
| AF110396.1 | - | 6 - 7 | 731 | 82 | ||
| AF112979.1 | - | 6 - 7 | 731 | 82 | ||
| AK032221.1 | - | 7 - 8 | 907 | 82 | ||
| AK052317.1 | - | 8 - 9 | 968 | 82 | ||
| AK083257.1 | - | 7 - 8 | 920 | 82 | ||
| AK145091.1 | - | 6 - 7 | 648 | 82 | ||
| AK145624.1 | - | 7 - 8 | 927 | 82 | ||
| AK156660.1 | - | 6 - 7 | 864 | 82 | ||
| AK220183.1 | - | 7 - 8 | 942 | 82 | ||
| BC003989.1 | - | 6 - 7 | 737 | 3491843 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Btrc
Genomic Map