Assay Details
| Gene Symbol: Mapkapk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010765.2 | NP_034895.1 | 2 - 3 | 793 | 66 | |
| GenBank mRNA | AF039840.1 | - | 2 - 3 | 807 | 66 | |
| AY533679.1 | - | 2 - 3 | 101 | 66 | ||
| AY533680.2 | - | 2 - 3 | 101 | 66 | ||
| BC019184.1 | - | 2 - 3 | 215 | 5010305 | 66 | |
| BY722898.1 | - | 2 - 3 | 378 | 66 | ||
More Information
Set Membership: |
|

Mapkapk5
Genomic Map