Assay Details
| Gene Symbol: Ptpn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109992.1 | NP_001103462.1 | 2 - 3 | 318 | 81 | |
| NM_011202.3 | NP_035332.1 | 2 - 3 | 318 | 81 | ||
| GenBank mRNA | AK011468.1 | - | 2 - 3 | 280 | 81 | |
| AK147354.1 | - | 2 - 3 | 289 | 81 | ||
| AK159501.1 | - | 2 - 3 | 318 | 81 | ||
| AK159587.1 | - | 2 - 3 | 318 | 81 | ||
| BC028513.1 | - | 2 - 3 | 268 | 81 | ||
| BC057398.1 | - | 2 - 3 | 250 | 6409640 | 81 | |
| BC059278.1 | - | 2 - 3 | 296 | 2866038 | 81 | |
| D84372.1 | - | 2 - 3 | 194 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn11
Genomic Map