Assay Details
| Gene Symbol: Ptpn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109992.1 | NP_001103462.1 | 9 - 10 | 1275 | 104 | |
| NM_011202.3 | NP_035332.1 | 9 - 10 | 1275 | 104 | ||
| GenBank mRNA | AK147354.1 | - | 8 - 9 | 104 | ||
| AK159501.1 | - | 9 - 10 | 1275 | 104 | ||
| AK159587.1 | - | 9 - 10 | 1275 | 104 | ||
| AK163809.1 | - | 6 - 7 | 703 | 104 | ||
| BC057398.1 | - | 9 - 10 | 1207 | 6409640 | 104 | |
| BC059278.1 | - | 9 - 10 | 1253 | 2866038 | 104 | |
| D84372.1 | - | 9 - 10 | 1151 | 104 | ||
| S59187.1 | - | 2 - 3 | 193 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn11
Genomic Map