Assay Details
| Gene Symbol: Zdhhc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023740.2 | NP_076229.2 | 3 - 4 | 594 | 61 | |
| GenBank mRNA | AF176814.1 | - | 3 - 4 | 567 | 61 | |
| AK005255.1 | - | 1 - 2 | 190 | 61 | ||
| AK151505.1 | - | 3 - 4 | 576 | 61 | ||
| AK168042.1 | - | 3 - 4 | 569 | 61 | ||
| BC010835.1 | - | 3 - 4 | 558 | 4239913 | 61 | |
| CK619221.1 | - | 3 - 4 | 581 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc16
Genomic Map