Assay Details
| Gene Symbol: Asah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019734.2 | NP_062708.1 | 10 - 11 | 890 | 70 | |
| GenBank mRNA | AF157500.1 | - | 10 - 11 | 820 | 70 | |
| AF352179.1 | - | 10 - 11 | 832 | 70 | ||
| AK075666.1 | - | 10 - 11 | 805 | 70 | ||
| AK075919.1 | - | 10 - 11 | 844 | 70 | ||
| AK077721.1 | - | 10 - 11 | 843 | 70 | ||
| AK090314.1 | - | 10 - 11 | 840 | 70 | ||
| AK150567.1 | - | 10 - 11 | 802 | 70 | ||
| AK151208.1 | - | 10 - 11 | 891 | 70 | ||
| AK151312.1 | - | 10 - 11 | 802 | 70 | ||
| AK151478.1 | - | 10 - 11 | 881 | 70 | ||
| AK152305.1 | - | 10 - 11 | 842 | 70 | ||
| AK152811.1 | - | 10 - 11 | 802 | 70 | ||
| AK152920.1 | - | 10 - 11 | 850 | 70 | ||
| AK153101.1 | - | 10 - 11 | 850 | 70 | ||
| AK153297.1 | - | 10 - 11 | 804 | 70 | ||
| AK159556.1 | - | 10 - 11 | 812 | 70 | ||
| AK166757.1 | - | 10 - 11 | 822 | 70 | ||
| BC003204.1 | - | 10 - 11 | 810 | 3587042 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asah1
Genomic Map