Assay Details
| Gene Symbol: Zfyve27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164531.1 | NP_001158003.1 | 4 - 5 | 573 | 66 | |
| NM_177319.3 | NP_796293.2 | 4 - 5 | 690 | 66 | ||
| GenBank mRNA | AK159065.1 | - | 4 - 5 | 547 | 66 | |
| BC024801.1 | - | 4 - 5 | 664 | 66 | ||
| BC042595.1 | - | 4 - 5 | 636 | 5363403 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfyve27
Genomic Map