Assay Details
| Gene Symbol: Zfyve27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164531.1 | NP_001158003.1 | 7 - 8 | 918 | 124 | |
| NM_177319.3 | NP_796293.2 | 7 - 8 | 1035 | 124 | ||
| GenBank mRNA | AK159065.1 | - | 7 - 8 | 892 | 124 | |
| BC024801.1 | - | 7 - 8 | 1009 | 124 | ||
| BC042595.1 | - | 7 - 8 | 981 | 5363403 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfyve27
Genomic Map