Assay Details
| Gene Symbol: Zfyve27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164531.1 | NP_001158003.1 | 8 - 9 | 990 | 82 | |
| GenBank mRNA | AK159065.1 | - | 8 - 9 | 964 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfyve27
Genomic Map