Assay Details
| Gene Symbol: Slc7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044740.2 | NP_001038205.1 | 9 - 10 | 1710 | 76 | |
| NM_007514.3 | NP_031540.2 | 9 - 10 | 1707 | 76 | ||
| GenBank mRNA | AK154621.1 | - | 9 - 10 | 1684 | 76 | |
| AK155558.1 | - | 9 - 10 | 1686 | 76 | ||
| AK171369.1 | - | 9 - 10 | 1682 | 76 | ||
| BC021653.1 | - | 9 - 10 | 1702 | 76 | ||
| BC127082.1 | - | 9 - 10 | 1623 | 40111372 | 76 | |
| DQ086834.1 | - | 9 - 10 | 1662 | 76 | ||
| L03290.1 | - | 8 - 9 | 1509 | 76 | ||
| L11600.1 | - | 8 - 9 | 1509 | 76 | ||
| L29006.1 | - | 8 - 9 | 2032 | 76 | ||
| M62838.1 | - | 9 - 10 | 1623 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a2
Genomic Map