Assay Details
| Gene Symbol: Dr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026106.4 | NP_080382.2 | 1 - 2 | 918 | 72 | |
| GenBank mRNA | AK007230.1 | - | 1 - 2 | 893 | 72 | |
| AK018040.1 | - | 1 - 2 | 841 | 72 | ||
| AK031057.1 | - | 1 - 2 | 852 | 72 | ||
| AK075713.1 | - | 1 - 2 | 918 | 72 | ||
| AK090113.1 | - | 1 - 2 | 900 | 72 | ||
| AK139866.1 | - | 1 - 2 | 848 | 72 | ||
| AK144749.1 | - | 1 - 2 | 912 | 72 | ||
| AK146573.1 | - | 1 - 2 | 881 | 72 | ||
| AK178588.1 | - | 1 - 2 | 138 | 72 | ||
| BC013461.1 | - | 1 - 2 | 487 | 4222118 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dr1
Genomic Map