Assay Details
| Gene Symbol: Ffar4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181748.2 | NP_861413.1 | 1 - 2 | 613 | 86 | |
| GenBank mRNA | AB115769.1 | - | 1 - 2 | 565 | 86 | |
| AK131616.1 | - | 1 - 2 | 600 | 86 | ||
| AY288424.1 | - | 1 - 2 | 565 | 86 | ||
| BC053698.1 | - | 1 - 2 | 613 | 6529120 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ffar4
Genomic Map