Assay Details
| Gene Symbol: Fra10ac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081075.1 | NP_001074544.1 | 4 - 5 | 391 | 113 | |
| GenBank mRNA | AK017666.1 | - | 4 - 5 | 453 | 113 | |
| AK077572.1 | - | 4 - 5 | 364 | 113 | ||
| AK172241.1 | - | 4 - 5 | 398 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fra10ac1
Genomic Map