Assay Details
| Gene Symbol: Cnnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031396.2 | NP_113573.2 | 6 - 7 | 2188 | 54 | |
| GenBank mRNA | AF202994.1 | - | 6 - 7 | 1414 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnnm1
Genomic Map