Assay Details
| Gene Symbol: P2rx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038839.2 | NP_001033928.1 | 3 - 4 | 525 | 76 | |
| NM_001038845.2 | NP_001033934.1 | 3 - 4 | 525 | 76 | ||
| NM_001284402.1 | NP_001271331.1 | 3 - 4 | 353 | 76 | ||
| NM_011027.3 | NP_035157.2 | 3 - 4 | 525 | 76 | ||
| GenBank mRNA | AJ009823.1 | - | 3 - 4 | 373 | 76 | |
| AJ489296.1 | - | 3 - 4 | 362 | 76 | ||
| AJ489297.1 | - | 3 - 4 | 362 | 76 | ||
| AK144585.1 | - | 3 - 4 | 526 | 76 | ||
| AK145050.1 | - | 3 - 4 | 380 | 76 | ||
| AK145211.1 | - | 3 - 4 | 389 | 76 | ||
| AK171132.1 | - | 3 - 4 | 497 | 76 | ||
| BC128303.1 | - | 3 - 4 | 369 | 76 | ||
| BC128304.1 | - | 3 - 4 | 369 | 76 | ||
| BC141120.1 | - | 3 - 4 | 392 | 9055732 | 76 | |
| BY752747.1 | - | 3 - 4 | 467 | 76 | ||
| FJ436444.1 | - | 3 - 4 | 353 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P2rx7
Genomic Map