Assay Details
| Gene Symbol: P2rx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038839.2 | NP_001033928.1 | 8 - 9 | 1041 | 78 | |
| NM_001038845.2 | NP_001033934.1 | 8 - 9 | 1041 | 78 | ||
| NM_001284402.1 | NP_001271331.1 | 8 - 9 | 869 | 78 | ||
| NM_011027.3 | NP_035157.2 | 8 - 9 | 1041 | 78 | ||
| GenBank mRNA | AJ489296.1 | - | 8 - 9 | 878 | 78 | |
| AJ489297.1 | - | 8 - 9 | 878 | 78 | ||
| AK144585.1 | - | 8 - 9 | 1042 | 78 | ||
| AK171132.1 | - | 8 - 9 | 1013 | 78 | ||
| BC128303.1 | - | 8 - 9 | 885 | 78 | ||
| BC128304.1 | - | 8 - 9 | 885 | 78 | ||
| BC141120.1 | - | 8 - 9 | 908 | 9055732 | 78 | |
| FJ436444.1 | - | 8 - 9 | 869 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P2rx7
Genomic Map