Assay Details
| Gene Symbol: Clcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177770.1 | NP_001171241.1 | 3 - 4 | 597 | 165 | |
| NM_001177771.1 | NP_001171242.1 | 4 - 5 | 512 | 165 | ||
| NM_145543.2 | NP_663518.1 | 3 - 4 | 309 | 165 | ||
| GenBank mRNA | AK090092.1 | - | 3 - 4 | 283 | 165 | |
| AK143631.1 | - | 4 - 5 | 495 | 165 | ||
| AK169803.1 | - | 3 - 4 | 307 | 165 | ||
| AK171296.1 | - | 3 - 4 | 336 | 165 | ||
| BC003247.1 | - | 3 - 4 | 340 | 3498611 | 165 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clcc1
Genomic Map