Assay Details
| Gene Symbol: Henmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078646.1 | NP_001072114.1 | 4 - 5 | 421 | 136 | |
| NM_025723.2 | NP_079999.2 | 4 - 5 | 736 | 136 | ||
| GenBank mRNA | AK038994.1 | - | 4 - 5 | 422 | 136 | |
| AK085587.1 | - | 4 - 5 | 736 | 136 | ||
| BC099408.1 | - | 3 - 4 | 402 | 30935249 | 136 | |
| BC116412.1 | - | 2 - 3 | 217 | 136 | ||
| BC116413.1 | - | 2 - 3 | 217 | 136 | ||
| BC128494.1 | - | 2 - 3 | 217 | 136 | ||
| BC128495.1 | - | 2 - 3 | 217 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Henmt1
Genomic Map