Assay Details
| Gene Symbol: Henmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078646.1 | NP_001072114.1 | 6 - 7 | 741 | 71 | |
| NM_025723.2 | NP_079999.2 | 6 - 7 | 1056 | 71 | ||
| GenBank mRNA | AK014900.1 | - | 6 - 7 | 1022 | 71 | |
| AK038994.1 | - | 6 - 7 | 742 | 71 | ||
| AK085587.1 | - | 6 - 7 | 1056 | 71 | ||
| BC099408.1 | - | 5 - 6 | 722 | 30935249 | 71 | |
| BC116412.1 | - | 4 - 5 | 415 | 71 | ||
| BC116413.1 | - | 4 - 5 | 537 | 71 | ||
| BC128494.1 | - | 4 - 5 | 537 | 71 | ||
| BC128495.1 | - | 4 - 5 | 537 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Henmt1
Genomic Map