Assay Details
| Gene Symbol: Myl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010861.3 | NP_034991.3 | 1 - 2 | 53 | 58 | |
| GenBank mRNA | AK002367.1 | - | 1 - 2 | 54 | 58 | |
| AK146674.1 | - | 1 - 2 | 57 | 58 | ||
| BC061144.1 | - | 54 | 30301284 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002367.1, AK146674.1, BC061144.1, NM_010861.3 transcripts.
Set Membership: |
|

Myl2
Genomic Map