Assay Details
| Gene Symbol: Ccdc63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183307.4 | NP_899130.1 | 1 - 2 | 202 | 112 | |
| GenBank mRNA | AK029513.1 | - | 1 - 2 | 203 | 113 | |
| BB614558.1 | - | 1 - 2 | 203 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029513.1, NM_183307.4 transcripts.
Set Membership: |
|

Ccdc63
Genomic Map