Assay Details
| Gene Symbol: Atxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009125.2 | NP_033151.2 | 14 - 15 | 2351 | 151 | |
| GenBank mRNA | AF041472.1 | - | 14 - 15 | 2350 | 151 | |
| AK135879.1 | - | 4 - 5 | 512 | 151 | ||
| AK135976.1 | - | 9 - 10 | 1308 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atxn2
Genomic Map