Assay Details
| Gene Symbol: Cept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293694.1 | NP_001280623.1 | 4 - 5 | 832 | 109 | |
| GenBank mRNA | BC021753.1 | - | 4 - 5 | 761 | 4167222 | 109 |
More Information
Updated Mapping Information
Set Membership: |
|

Cept1
Genomic Map