Assay Details
| Gene Symbol: Ntng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163350.1 | NP_001156822.1 | 7 - 8 | 2308 | 101 | |
| GenBank mRNA | AB038664.1 | - | 6 - 7 | 1213 | 101 | |
| AF475072.1 | - | 6 - 7 | 1213 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ntng1
Genomic Map