Assay Details
| Gene Symbol: Gtf2e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167922.1 | NP_001161394.1 | 1 - 2 | 171 | 96 | |
| GenBank mRNA | BX516160.1 | - | 1 - 2 | 308 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001167922.1 transcripts.
Set Membership: |
|

Gtf2e2
Genomic Map